missing translation for 'onlineSavingsMsg'
Learn More

L-Lactic Acid 85.0+%, TCI America™

Catalog No. p-7131853
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L016525G 25 g
L0165500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L016525G Supplier TCI America Supplier No. L016525G
Only null left

Chemical Identifiers

CAS 79-33-4
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.078
MDL Number MFCD00064266
InChI Key JVTAAEKCZFNVCJ-REOHCLBHSA-N
Synonym l-lactic acid, l-+-lactic acid, s-2-hydroxypropanoic acid, s-lactic acid, sarcolactic acid, s-2-hydroxypropionic acid, 2s-2-hydroxypropanoic acid, +-lactic acid, espiritin, tisulac
PubChem CID 107689
ChEBI CHEBI:422
IUPAC Name (2S)-2-hydroxypropanoic acid
SMILES CC(C(=O)O)O

Specifications

Color Colorless
UN Number 3265
Formula Weight 90.08
Percent Purity ≥85.0% (T)
Quantity 25 g
Physical Form Liquid
Chemical Name or Material L-Lactic Acid
EINECSNumber (2)-1369
RTECSNumber OD3100000
TSCA Yes