missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(+)-Lactic acid, 90% solution in water

Catalog No. p-3598398
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC189872500 250 mL Glass bottle
AC189870010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC189872500 Supplier Thermo Scientific Chemicals Supplier No. 189872500
Only null left

CAS: 79-33-4 | C3H6O3 | 90.078 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 79-33-4
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.078
MDL Number MFCD00064266
InChI Key JVTAAEKCZFNVCJ-REOHCLBHSA-N
Synonym l-lactic acid, l-+-lactic acid, s-2-hydroxypropanoic acid, s-lactic acid, sarcolactic acid, s-2-hydroxypropionic acid, 2s-2-hydroxypropanoic acid, +-lactic acid, espiritin, tisulac
PubChem CID 107689
ChEBI CHEBI:422
IUPAC Name (2S)-2-hydroxypropanoic acid
SMILES CC(C(=O)O)O

Specifications

Boiling Point 125.0°C
Color Colorless to Yellow
Linear Formula CH3CH(OH)CO2H
Beilstein 03, IV, 633
Fieser 15,181
Merck Index 15, 5385
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, furfurol and ether, practically insoluble in chloroform and carbon, disulfide
Formula Weight 90.08
Percent Purity ≥88%
Density 1.2000g/mL
Quantity 250 mL
Flash Point >112°C
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.4260 to 1.4280
Specific Gravity 1.2
Chemical Name or Material L(+)-Lactic acid
Show More Show Less
Hazard Category Danger
Hazard Statement GHS P Statement
Causes serious eye damage.
Causes skin irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only