missing translation for 'onlineSavingsMsg'
Learn More

L-Isoleucine 98.0+%, TCI America™

Catalog No. p-7131257
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I018125G 25 g
I0181100G 100 g
I0181500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I018125G Supplier TCI America Supplier No. I018125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 73-32-5
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00064222 MFCD00004268
InChI Key AGPKZVBTJJNPAG-WHFBIAKZSA-N
Synonym l-isoleucine, isoleucine, 2s,3s-2-amino-3-methylpentanoic acid, s-isoleucine, s,s-isoleucine, 2s,3s-isoleucine, 2-amino-3-methylvaleric acid, erythro-l-isoleucine, l-+-isoleucine, l-ile
PubChem CID 6306
ChEBI CHEBI:17191
IUPAC Name (2S,3S)-2-amino-3-methylpentanoic acid
SMILES CC[C@H](C)[C@H](N)C(O)=O

Specifications

Melting Point 288°C
Color White
Quantity 25 g
Formula Weight 131.18
Percent Purity ≥98.0% (T)
Grade Laboratory
Physical Form Crystalline Powder
Chemical Name or Material L-Isoleucine
EINECSNumber (9)-1560
RTECSNumber NR4705000
TSCA Yes