missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Isoleucine, Multi-Compendial, U.S.P., J.T. Baker™
$4,466.00 - $25,184.78
Chemical Identifiers
| CAS | 73-32-5 |
|---|---|
| Molecular Formula | C6H13NO2 |
| Molecular Weight (g/mol) | 131.18 |
| MDL Number | MFCD00064222 MFCD00004268 |
| InChI Key | AGPKZVBTJJNPAG-WHFBIAKZSA-N |
| Synonym | l-isoleucine, isoleucine, 2s,3s-2-amino-3-methylpentanoic acid, s-isoleucine, s,s-isoleucine, 2s,3s-isoleucine, 2-amino-3-methylvaleric acid, erythro-l-isoleucine, l-+-isoleucine, l-ile |
| PubChem CID | 6306 |
| ChEBI | CHEBI:17191 |
| IUPAC Name | (2S,3S)-2-amino-3-methylpentanoic acid |
| SMILES | CC[C@H](C)[C@H](N)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
02003206
|
Avantor J.T.Baker
208205 |
100 g | Poly Bottle |
Each for $4,466.00
|
|
||||
|
02003208
|
Avantor J.T.Baker
208206 |
1 kg | Poly Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| C6H13NO2 | |
| MFCD00064222 MFCD00004268 | |
| l-isoleucine, isoleucine, 2s,3s-2-amino-3-methylpentanoic acid, s-isoleucine, s,s-isoleucine, 2s,3s-isoleucine, 2-amino-3-methylvaleric acid, erythro-l-isoleucine, l-+-isoleucine, l-ile | |
| CHEBI:17191 | |
| CC[C@H](C)[C@H](N)C(O)=O |
Specifications
| 73-32-5 | |
| MFCD00064222 MFCD00004268 | |
| AGPKZVBTJJNPAG-WHFBIAKZSA-N | |
| (2S,3S)-2-amino-3-methylpentanoic acid | |
| 6306 | |
| 131.17 | |
| L-Isoleucine |
| C6H13NO2 | |
| l-isoleucine, isoleucine, 2s,3s-2-amino-3-methylpentanoic acid, s-isoleucine, s,s-isoleucine, 2s,3s-isoleucine, 2-amino-3-methylvaleric acid, erythro-l-isoleucine, l-+-isoleucine, l-ile | |
| CC[C@H](C)[C@H](N)C(O)=O | |
| 131.18 | |
| CHEBI:17191 | |
| Multi-Compendial/USP |