missing translation for 'onlineSavingsMsg'
Learn More

L-Isoleucine methyl ester hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog No. AAA1784706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1784706 5 g
AAA1784714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1784706 Supplier Thermo Scientific Chemicals Supplier No. A1784706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

L-Isoleucine methyl ester used for chemical synthesis. E.g. It can also be used to produce 3-methyl-2-(4-nitro-imidazol-1-yl)-pentanoic acid methyl ester. It also finds its use as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Isoleucine methyl ester used for chemical synthesis. E.g. It can also be used to produce 3-methyl-2-(4-nitro-imidazol-1-yl)-pentanoic acid methyl ester. It also finds its use as a pharmaceutical intermediate.

Solubility
Soluble in methanol (50 mg/ml clear, colorless solution).

Notes
Hygroscopic. Keep away from oxidizing agents, water/moisture. Store under inert atmosphere. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. In order to obtain a higher solubility, warm the tube at 37°C and shake it in the ultrasonic bath for a while.

Chemical Identifiers

CAS 18598-74-8
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
MDL Number MFCD00038911
InChI Key GGTBEWGOPAFTTH-GEMLJDPKSA-N
Synonym l-isoleucine methyl ester hydrochloride, h-ile-ome.hcl, methyl l-isoleucinate hydrochloride, methyl l-isoleucinate hcl, methyl 2s,3s-2-amino-3-methylpentanoate hydrochloride, 2s,3s-methyl 2-amino-3-methylpentanoate hydrochloride, l-isoleucine, methyl ester, hydrochloride, l-isoleucine methyl ester hydrochloride;h-ile-ome.hcl, h-lle-ome.hcl, h-lle-ome?cl
PubChem CID 11171351
IUPAC Name methyl (2S,3S)-2-amino-3-methylpentanoate;hydrochloride
SMILES Cl.CC[C@H](C)[C@H](N)C(=O)OC

Specifications

Melting Point 98°C to 100°C
Beilstein 3595132
Solubility Information Soluble in methanol (50mg/ml clear,colorless solution).
Quantity 5 g
Formula Weight 181.66
Sensitivity Hygroscopic
Percent Purity ≥98%
Chemical Name or Material L-Isoleucine methyl ester hydrochloride
EINECSNumber 242-437-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only