missing translation for 'onlineSavingsMsg'
Learn More

L-Isoleucine, >99%, MP Biomedicals™

Catalog No. ICN10209291
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10209291 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10209291 Supplier MP Biomedicals Inc Supplier No. 0210209291

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 73-32-5
Color White
Assay Percent Range >99%
Molecular Formula C6H13NO2
MDL Number MFCD00064222 MFCD00004268
Synonym l-isoleucine, isoleucine, 2s,3s-2-amino-3-methylpentanoic acid, s-isoleucine, s,s-isoleucine, 2s,3s-isoleucine, 2-amino-3-methylvaleric acid, erythro-l-isoleucine, l-+-isoleucine, l-ile
InChI Key AGPKZVBTJJNPAG-WHFBIAKZSA-N
SMILES CC[C@H](C)[C@H](N)C(O)=O
IUPAC Name (2S,3S)-2-amino-3-methylpentanoic acid
Molecular Weight (g/mol) 131.18
Quantity 1 kg
PubChem CID 6306
ChEBI CHEBI:17191
Formula Weight 131.2
Percent Purity 1.003
Physical Form Powder
Chemical Name or Material L-Isoleucine
Show More Show Less
EINECSNumber 200-798-2
Recommended Storage Store at Room Temperature (15 to 30°C).