missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals L-Isoleucine, 99%
CAS: 73-32-5 | C6H13NO2 | 131.18 g/mol
Supplier: Thermo Scientific Chemicals 166171000
| Quantity | 100 g |
|---|---|
| Packaging | Plastic Bottle |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C6H13NO2 | |
| MFCD00064222 MFCD00004268 | |
| l-isoleucine, isoleucine, 2s,3s-2-amino-3-methylpentanoic acid, s-isoleucine, s,s-isoleucine, 2s,3s-isoleucine, 2-amino-3-methylvaleric acid, erythro-l-isoleucine, l-+-isoleucine, l-ile | |
| CHEBI:17191 | |
| CC[C@H](C)[C@H](N)C(O)=O |
Specifications
| 284.0°C to 286.0°C | |
| 73-32-5 | |
| 500ppm max. | |
| Authentic | |
| 0.3% max. (105°C, 3 hrs) | |
| Plastic Bottle | |
| C2H5CH(CH3)CH(NH2)CO2H | |
| 0.3% max. | |
| 04, 454 | |
| 15, 5225 | |
| Solubility in water: 41.2g/L (50°C). Other solubilities: soluble in mineral acids, practically insoluble in alcohol, insoluble in ether | |
| AGPKZVBTJJNPAG-WHFBIAKZSA-N | |
| (2S,3S)-2-amino-3-methylpentanoic acid | |
| 131.18 | |
| 6306 | |
| 131.17 | |
| Crystalline Powder or Crystals |
| L-Isoleucine | |
| White | |
| 20ppm max. | |
| 30ppm max. | |
| 99% | |
| C6H13NO2 | |
| MFCD00064222 MFCD00004268 | |
| + 41.00 | |
| 300ppm max. | |
| l-isoleucine, isoleucine, 2s,3s-2-amino-3-methylpentanoic acid, s-isoleucine, s,s-isoleucine, 2s,3s-isoleucine, 2-amino-3-methylvaleric acid, erythro-l-isoleucine, l-+-isoleucine, l-ile | |
| 168.0 °C | |
| CC[C@H](C)[C@H](N)C(O)=O | |
| +41° (20°C c=4,6N HCl) | |
| 100 g | |
| CHEBI:17191 | |
| 99% |