missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Isoleucinamide Hydrochloride 98.0+%, TCI America™
$46.60 - $128.62
Chemical Identifiers
| CAS | 10466-56-5 |
|---|---|
| Molecular Formula | C6H15ClN2O |
| Molecular Weight (g/mol) | 166.649 |
| MDL Number | MFCD00058476 |
| InChI Key | SAOZPTBFWAEJQL-FHAQVOQBSA-N |
| PubChem CID | 16219521 |
| IUPAC Name | (2S,3S)-2-amino-3-methylpentanamide;hydrochloride |
| SMILES | CCC(C)C(C(=O)N)N.Cl |
Chemical Identifiers
| 10466-56-5 | |
| 166.649 | |
| SAOZPTBFWAEJQL-FHAQVOQBSA-N | |
| (2S,3S)-2-amino-3-methylpentanamide;hydrochloride |
| C6H15ClN2O | |
| MFCD00058476 | |
| 16219521 | |
| CCC(C)C(C(=O)N)N.Cl |
Specifications
| 10466-56-5 | |
| White | |
| MFCD00058476 | |
| SAOZPTBFWAEJQL-FHAQVOQBSA-N | |
| (2S,3S)-2-amino-3-methylpentanamide;hydrochloride | |
| 16219521 | |
| ≥98.0% (N) | |
| L-Isoleucinamide Hydrochloride |
| 246°C | |
| C6H15ClN2O | |
| 1 g | |
| CCC(C)C(C(=O)N)N.Cl | |
| 166.649 | |
| 166.65 | |
| Crystalline Powder |
Safety and Handling
TSCA : No