missing translation for 'onlineSavingsMsg'
Learn More

L-Isoleucinamide Hydrochloride 98.0+%, TCI America™

Catalog No. I07311G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07311G 1 g
I07315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I07311G Supplier TCI America Supplier No. I07311G
Only null left

Chemical Identifiers

CAS 10466-56-5
Molecular Formula C6H15ClN2O
Molecular Weight (g/mol) 166.649
MDL Number MFCD00058476
InChI Key SAOZPTBFWAEJQL-FHAQVOQBSA-N
PubChem CID 16219521
IUPAC Name (2S,3S)-2-amino-3-methylpentanamide;hydrochloride
SMILES CCC(C)C(C(=O)N)N.Cl

Specifications

Melting Point 246°C
Color White
Quantity 1 g
Formula Weight 166.65
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material L-Isoleucinamide Hydrochloride