missing translation for 'onlineSavingsMsg'
Learn More

L(-)-Histidinol dihydrochloride, 99%

Catalog No. AC228830010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC228830010 1 g
AC228830050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC228830010 Supplier Thermo Scientific Chemicals Supplier No. 228830010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1596-64-1 | C6H13Cl2N3O | 214.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1596-64-1
Molecular Formula C6H13Cl2N3O
Molecular Weight (g/mol) 214.09
MDL Number MFCD00078058
InChI Key FRCAFNBBXRWXQA-XRIGFGBMSA-N
Synonym l-histidinol dihydrochloride, l---histidinol dihydrochloride, beta-aminoimidazole-4-propanol dihydrochloride, l-histidinol 2 hcl, s-2-amino-3-4-imidazolyl propanol dihydrochloride, l--histidinol 2hcl, imidazole-4-propanol, beta-amino-, dihydrochloride, s-beta-amino-1h-imidazole-4-propanol dihydrochloride, s-2-amino-3-1h-imidazol-4-yl propan-1-ol dihydrochloride, 1h-imidazole-4-propanol, beta-amino-, dihydrochloride, s-9ci
PubChem CID 197743
IUPAC Name (2S)-2-amino-3-(1H-imidazol-5-yl)propan-1-ol;dihydrochloride
SMILES C1=C(NC=N1)CC(CO)N.Cl.Cl

Specifications

Infrared Spectrum Authentic
Color White to Beige
Packaging Glass bottle
Specific Rotation − 3.00
Specific Rotation Condition −3° (24°C c=2,H2O)
Melting Point 193.0°C to 199.0°C
Quantity 1 g
Physical Form Crystalline Powder
Assay Percent Range 99%
Formula Weight 214.1
Percent Purity 99%
Chemical Name or Material L(-)-Histidinol dihydrochloride
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only