missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Gulonic acid gamma-lactone, 95+%

Catalog No. AC255570250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC255570250 25 g Glass bottle
AC255571000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC255570250 Supplier Thermo Scientific Chemicals Supplier No. 255570250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1128-23-0 | C6H10O6 | 178.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1128-23-0
Molecular Formula C6H10O6
Molecular Weight (g/mol) 178.14
InChI Key SXZYCXMUPBBULW-SKNVOMKLSA-N
Synonym l-gulonolactone, l-gulono-1,4-lactone, l-gulonic acid gamma-lactone, 3s,4r,5r-5-s-1,2-dihydroxyethyl-3,4-dihydroxydihydrofuran-2 3h-one, l-gulono-gamma-lactone, gamma-gulonolactone, l-gulonic gamma-lactone, l-+-gulono-1,4-lactone, l +-gulonic acid gamma-lactone, l-+-gulonic acid gamma-lactone
PubChem CID 439373
ChEBI CHEBI:17587
IUPAC Name (3S,4R,5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxyoxolan-2-one
SMILES C(C(C1C(C(C(=O)O1)O)O)O)O

Specifications

Melting Point 183.0°C to 185.0°C
Loss on Drying 0.5% max. (1 g, 105°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 95% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 25 g
Specific Rotation Condition + 54.50 (57.50°C c=2,water)
Specific Rotation + 54.50
Formula Weight 178.14
Percent Purity 95+%
Physical Form Viscous Solution
Chemical Name or Material L(+)-Gulonic acid γ-lactone
Show More Show Less

RUO – Research Use Only