missing translation for 'onlineSavingsMsg'
Learn More

L-glycero-alpha-D-manno-Heptopyranose 1,2,3,4,6,7-Hexaacetate 98.0+%, TCI America™

Catalog No. H165720MG
Change view
Click to view available options
Quantity:
20 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H165720MG 20 mg
H1657100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H165720MG Supplier TCI America Supplier No. H165720MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 104486-80-8
Molecular Formula C19H26O13
Synonym LD-Heptose Peracetate

Specifications

Melting Point 133°C
Color White
Quantity 20 mg
Formula Weight 462.40
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material L-glycero-alpha-D-manno-Heptopyranose 1,2,3,4,6,7-Hexaacetate
TSCA No
Recommended Storage Refrigerator