Learn More
L-Glutathione, reduced, 98+%
A tripeptide used for reductive cleavage of disulfide bonds | CAS: 70-18-8 | C10H17N3O6S | 307.321 g/mol
Supplier: Thermo Scientific Chemicals J6216622
Description
A tripeptide used for the reductive cleavage of disulfide bonds L-Glutathione is used in the treatment of lung diseases for patients who are HIV positive. It protects the cancerous cells by conferring resistance to chemotherapeutic drugs. It is involved in many aspects of metabolism including transport of g-glutanyl amino acids and reductive cleavage of disulfide bonds. As an antioxidant, it prevents damage to important cellular components that arise due to reactive oxygen species like free radicals and peroxide. It is also used to decrease the concentrations of inflammatory cytokines (IL-6, IL-18) as well as involved in increasing the level of serum Ca2+ ions. It is also used in white wine production.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in water, dimethyl formamide, ethanol, phosphate buffered saline, diluted alcohol, liquid ammonia and dimethyl sulfoxide.
Notes
Store in cool place. Incompatible with strong oxidizing agents.
Specifications
| L-Glutathione | |
| 70-18-8 | |
| White | |
| MFCD00065939 | |
| 1729812 | |
| glutathione, l-glutathione, glutathion, glutathione-sh, glutinal, isethion, tathion, reduced glutathione, deltathione, neuthion | |
| RWSXRVCMGQZWBV-WDSKDSINSA-N | |
| C(CC(=O)NC(CS)C(=O)NCC(=O)O)C(C(=O)O)N | |
| 307.321 | |
| CHEBI:16856 | |
| ≥98% |
| reduced | |
| ∽193°C (decomposition) | |
| C10H17N3O6S | |
| 100 g | |
| 14,4475 | |
| Soluble in water,dimethyl formamide,ethanol,phosphate buffered saline,diluted alcohol,liquid ammonia and dimethyl sulfoxide. | |
| −17° (c=2 in Water) | |
| (2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-1-oxo-3-sulfanylpropan-2-yl]amino]-5-oxopentanoic acid | |
| 124886 | |
| 307.33 | |
| Crystalline Powder |
Chemical Identifiers
| 70-18-8 | |
| 307.321 | |
| RWSXRVCMGQZWBV-WDSKDSINSA-N | |
| 124886 | |
| (2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-1-oxo-3-sulfanylpropan-2-yl]amino]-5-oxopentanoic acid |
| C10H17N3O6S | |
| MFCD00065939 | |
| glutathione, l-glutathione, glutathion, glutathione-sh, glutinal, isethion, tathion, reduced glutathione, deltathione, neuthion | |
| CHEBI:16856 | |
| C(CC(=O)NC(CS)C(=O)NCC(=O)O)C(C(=O)O)N |
Safety and Handling
EINECSNumber : 200-725-4
RTECSNumber : MC0556000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only