missing translation for 'onlineSavingsMsg'
Learn More

L-Glutathione, Reduced, 98.0-101.0%, Spectrum™ Chemical

Catalog No. p-7165624
Change view
Click to view available options
Quantity:
25 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611276 25 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18611276 Supplier Spectrum Chemical Mfg Cor Supplier No. GL14625GM
Only null left

Chemical Identifiers

CAS 70-18-8
Molecular Formula C10H17N3O6S
Molecular Weight (g/mol) 307.32
InChI Key RWSXRVCMGQZWBV-UHFFFAOYNA-N
IUPAC Name 2-amino-4-({1-[(carboxymethyl)carbamoyl]-2-sulfanylethyl}carbamoyl)butanoic acid
SMILES NC(CCC(=O)NC(CS)C(=O)NCC(O)=O)C(O)=O

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.005
Packaging Amber Glass Bottle
Quantity 25 g
Specific Rotation -17.5° to -15.5°
Percent Purity 98 to 101%
Grade Reagent

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.