missing translation for 'onlineSavingsMsg'
Learn More

L-Glutathione, oxidized

Acts as hydrogen acceptor in enzymatic determination of NADP and NADPH | CAS: 27025-41-8 | C20H32N6O12S2 | 612.63 g/mol

Supplier:  Thermo Scientific Chemicals J6371506

 View more versions of this product

Catalog No. AAJ6371506


Only null left
Explore available promotions

Description

Description

L-Glutathione is used as a Acts as hydrogen acceptor in enzymatic determination of NADP and NADPH, it is used to reduced back to glutathione (GSH) via the NADPH-dependent enzyme glutathione reductase. The ratio of GSH to GSSG is often used to gauge the level of oxidative stress in cells, with higher concentrations of GSSG implying more oxidative stress

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Glutathione is used as a Acts as hydrogen acceptor in enzymatic determination of NADP and NADPH, it is used to reduced back to glutathione (GSH) via the NADPH-dependent enzyme glutathione reductase. The ratio of GSH to GSSG is often used to gauge the level of oxidative stress in cells, with higher concentrations of GSSG implying more oxidative stress

Solubility
Soluble in water, DMF, and ethanol.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

L-Glutathione, oxidized
White
MFCD00150701
1718700
oxiglutatione, glutathione disulfide, gssg, oxidized glutathione, glutathione disulphide, glutathione-ssg, l-glutathione oxidized, oxigluthione, glutathone disulfide, glutathione oxidized
YPZRWBKMTBYPTK-IOKZUGQQNA-N
N[C@@H](CCC(=O)N[C@@H](CSSC[C@H](NC(=O)CC[C@H](N)C(O)=O)C(=O)NCC(O)=O)C(=O)NCC(O)=O)C(O)=O
612.63
CHEBI:17858
Powder
27025-41-8
C20H32N6O12S2
5 g
14,4475
Soluble in water,DMF,and ethanol.
−105° (c=2 in Water)
(2S)-2-amino-5-[[(2R)-3-[[(2R)-2-[[(4S)-4-amino-4-carboxybutanoyl]amino]-3-(carboxymethylamino)-3-oxopropyl]disulfanyl]-1-(carboxymethylamino)-1-oxopropan-2-yl]amino]-5-oxopentanoic acid
65359
612.64

Chemical Identifiers

27025-41-8
612.63
YPZRWBKMTBYPTK-IOKZUGQQNA-N
65359
(2S)-2-amino-5-[[(2R)-3-[[(2R)-2-[[(4S)-4-amino-4-carboxybutanoyl]amino]-3-(carboxymethylamino)-3-oxopropyl]disulfanyl]-1-(carboxymethylamino)-1-oxopropan-2-yl]amino]-5-oxopentanoic acid
C20H32N6O12S2
MFCD00150701
oxiglutatione, glutathione disulfide, gssg, oxidized glutathione, glutathione disulphide, glutathione-ssg, l-glutathione oxidized, oxigluthione, glutathone disulfide, glutathione oxidized
CHEBI:17858
N[C@@H](CCC(=O)N[C@@H](CSSC[C@H](NC(=O)CC[C@H](N)C(O)=O)C(=O)NCC(O)=O)C(=O)NCC(O)=O)C(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 248-170-7

RTECSNumber : MC0556500

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only