missing translation for 'onlineSavingsMsg'
Learn More

L(-)-Glutathione, oxidized, 98%

CAS: 27025-41-8 | C20H32N6O12S2 | 612.63 g/mol

Supplier:  Thermo Scientific Chemicals 320220050

 View more versions of this product

Catalog No. AC320220050


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

L(-)-Glutathione, oxidized
27025-41-8
White
Authentic
Glass bottle
MFCD00150701
04, IV, 3168
−99.00 (20.00°C c=4,water)
oxiglutatione, glutathione disulfide, gssg, oxidized glutathione, glutathione disulphide, glutathione-ssg, l-glutathione oxidized, oxigluthione, glutathone disulfide, glutathione oxidized
YPZRWBKMTBYPTK-IOKZUGQQNA-N
2-amino-4-[(2-{[2-(4-amino-4-carboxybutanamido)-2-[(carboxymethyl)carbamoyl]ethyl]disulfanyl}-1-[(carboxymethyl)carbamoyl]ethyl)carbamoyl]butanoic acid
65359
612.64
Powder
98%
178.0°C
0.5% max. (800°C)
97.5% min. (HPLC)
C20H32N6O12S2
5 g
15, 4511
− 99.00
Solubility in water: soluble.
N[C@@H](CCC(=O)N[C@@H](CSSC[C@H](NC(=O)CC[C@H](N)C(O)=O)C(=O)NCC(O)=O)C(=O)NCC(O)=O)C(O)=O
612.63
CHEBI:17858
98%

Chemical Identifiers

27025-41-8
612.63
YPZRWBKMTBYPTK-IOKZUGQQNA-N
65359
2-amino-4-[(2-{[2-(4-amino-4-carboxybutanamido)-2-[(carboxymethyl)carbamoyl]ethyl]disulfanyl}-1-[(carboxymethyl)carbamoyl]ethyl)carbamoyl]butanoic acid
C20H32N6O12S2
MFCD00150701
oxiglutatione, glutathione disulfide, gssg, oxidized glutathione, glutathione disulphide, glutathione-ssg, l-glutathione oxidized, oxigluthione, glutathone disulfide, glutathione oxidized
CHEBI:17858
N[C@@H](CCC(=O)N[C@@H](CSSC[C@H](NC(=O)CC[C@H](N)C(O)=O)C(=O)NCC(O)=O)C(=O)NCC(O)=O)C(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 248-170-7

RTECSNumber : MC0556500

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only