missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L(+)-Glutamine, specified according to the requirements of USP
CAS: 56-85-9 | C5H10N2O3 | 146.15 g/mol
$105.72 - $1,092.54
Chemical Identifiers
| CAS | 56-85-9 |
|---|---|
| Molecular Formula | C5H10N2O3 |
| Molecular Weight (g/mol) | 146.15 |
| MDL Number | MFCD00008044 |
| InChI Key | ZDXPYRJPNDTMRX-VKHMYHEASA-N |
| Synonym | l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid |
| PubChem CID | 5961 |
| ChEBI | CHEBI:18050 |
| IUPAC Name | (2S)-2,5-diamino-5-oxopentanoic acid |
| SMILES | N[C@@H](CCC(N)=O)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC386030500
|
N/A | 50 g |
Each for $105.72
|
|
|||||
|
AC386030010
|
N/A | 1 kg |
Each for $1,092.54
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C5H10N2O3 | |
| MFCD00008044 | |
| l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid | |
| CHEBI:18050 | |
| N[C@@H](CCC(N)=O)C(O)=O |
Specifications
| 185.0°C | |
| C5H10N2O3 | |
| MFCD00008044 | |
| Solubility in water: 35g/L (20°C). Other solubilities: practically insoluble in methanol, ethanol, ether,, benzene, acetone, ethyl acetate, chloroform, soluble in mineral acids, soluble in ethanol: 46 μg/100mL (25°C), soluble in methanol: 3.5 mg/100mL (2 | |
| N[C@@H](CCC(N)=O)C(O)=O | |
| 146.15 | |
| CHEBI:18050 | |
| L(+)-Glutamine |
| 56-85-9 | |
| H2NCOCH2CH2CH(NH2)CO2H | |
| l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid | |
| ZDXPYRJPNDTMRX-VKHMYHEASA-N | |
| (2S)-2,5-diamino-5-oxopentanoic acid | |
| 5961 | |
| 146.15 |
RUO – Research Use Only