missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamine 99.0+%, TCI America™

Catalog No. G006325G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G006325G 25 g
G0063100G 100 g
G0063500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. G006325G Supplier TCI America Supplier No. G006325G

Chemical Identifiers

CAS 56-85-9
Molecular Formula C5H10N2O3
Molecular Weight (g/mol) 146.15
MDL Number MFCD00008044
InChI Key ZDXPYRJPNDTMRX-VKHMYHEASA-N
Synonym l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid
PubChem CID 5961
ChEBI CHEBI:18050
IUPAC Name (2S)-2-amino-4-carbamoylbutanoic acid
SMILES N[C@@H](CCC(N)=O)C(O)=O

Specifications

Melting Point 186°C
Color White
Quantity 25 g
Formula Weight 146.15
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Glutamine
EINECSNumber (9)-1581
RTECSNumber MA2275100
TSCA Yes