missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamine, FCC, 98.5-101.5%, Spectrum™ Chemical

Catalog No. 18603835
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603835 25 g Amber Glass Bottle
18603836 100 g Amber Glass Bottle
18603837 1 kg Amber Glass Bottle
18603838 12 kg Poly Pail
18603839 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18603835 Supplier Spectrum Chemical Mfg Cor Supplier No. G103925GM

G1039, 56-85-9, C5H10N2O3

Spectrum™ Chemical L-Glutamine, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 56-85-9
Molecular Formula C5H10N2O3
Molecular Weight (g/mol) 146.15
MDL Number MFCD00008044
InChI Key ZDXPYRJPNDTMRX-VKHMYHEASA-N
IUPAC Name (2S)-2-amino-4-carbamoylbutanoic acid
SMILES N[C@@H](CCC(N)=O)C(O)=O

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 0.3%
Packaging Amber Glass Bottle
Quantity 25 g
Optical Rotation +6.3° to +7.3°
Percent Purity 98.5 to 101.5%
Grade FCC