Learn More
Thermo Scientific Chemicals L-Glutamine, Cell Culture Reagent
CAS: 56-85-9 | C5H10N2O3 | 146.15 g/mol
Supplier: Thermo Scientific Chemicals J60573A1
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| L-Glutamine | |
| 5.0 to 6.0 | |
| C5H10N2O3 | |
| 1723797 | |
| l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid | |
| N[C@@H](CCC(N)=O)C(O)=O | |
| 146.15 | |
| 5961 | |
| 146.14 | |
| Crystalline Powder |
| 56-85-9 | |
| White | |
| MFCD00008044 | |
| 14,4471 | |
| ZDXPYRJPNDTMRX-VKHMYHEASA-N | |
| (2S)-2,5-diamino-5-oxopentanoic acid | |
| 1 kg | |
| CHEBI:18050 | |
| Cell Culture Reagent |
Chemical Identifiers
| 56-85-9 | |
| 146.15 | |
| ZDXPYRJPNDTMRX-VKHMYHEASA-N | |
| 5961 | |
| (2S)-2,5-diamino-5-oxopentanoic acid |
| C5H10N2O3 | |
| MFCD00008044 | |
| l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid | |
| CHEBI:18050 | |
| N[C@@H](CCC(N)=O)C(O)=O |
Safety and Handling
P261-P264b-P271-P280g-P280i-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H320-H335-H336
EINECSNumber : 200-292-1
RTECSNumber : MA2275100
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only