missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Glutamine, Cell Culture Reagent

Catalog No. AAJ6057322
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6057322 100 g
AAJ6057314 25 g
AAJ60573A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6057322 Supplier Thermo Scientific Chemicals Supplier No. J6057322
Only null left

CAS: 56-85-9 | C5H10N2O3 | 146.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56-85-9
Molecular Formula C5H10N2O3
Molecular Weight (g/mol) 146.15
MDL Number MFCD00008044
InChI Key ZDXPYRJPNDTMRX-VKHMYHEASA-N
Synonym l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid
PubChem CID 5961
ChEBI CHEBI:18050
IUPAC Name (2S)-2,5-diamino-5-oxopentanoic acid
SMILES N[C@@H](CCC(N)=O)C(O)=O

Specifications

pH 5.0 to 6.0
Color White
Beilstein 1723797
Merck Index 14,4471
Quantity 100 g
Formula Weight 146.14
Grade Cell Culture Reagent
Physical Form Crystalline Powder
Chemical Name or Material L-Glutamine
Hazard Category H315-H319-H320-H335-H336
Precautionary Statement P261-P264b-P271-P280g-P280i-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 200-292-1
RTECSNumber MA2275100
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only