missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamine, ≥98.5%, Ultrapure

CAS: 56-85-9 | C5H10N2O3 | 146.146 g/mol

Supplier:  Thermo Fisher Scientific J1628536

Quantity 500 g
Packaging Plastic Bottle

 View more versions of this product

Catalog No. AAJ1628536


Only null left

Description

Description

Specifications

Chemical Identifiers

56-85-9
146.146
ZDXPYRJPNDTMRX-VKHMYHEASA-N
5961
(2 S)-2,5-diamino-5-oxopentanoic acid
C5H10N2O3
MFCD00008044
l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid
CHEBI:18050
C(CC(=O)N)C(C(=O)O)N

Specifications

L-Glutamine
Plastic Bottle
H2NCOCH2CH2CH(NH2)COOH
1723797
ZDXPYRJPNDTMRX-VKHMYHEASA-N
(2 S)-2,5-diamino-5-oxopentanoic acid
500 g
CHEBI:18050
≥98.5%
56-85-9
C5H10N2O3
MFCD00008044
l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid
C(CC(=O)N)C(C(=O)O)N
146.146
5961
146.14
Crystalline Powder
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280g-P280i-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H320-H335-H336

EINECSNumber : 200-292-1

RTECSNumber : MA2275100

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions