missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamine, 98.5-101.5%, Spectrum™ Chemical

Catalog No. p-7165623
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611271 25 g Amber Glass Bottle
18611270 100 g Amber Glass Bottle
18611269 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18611271 Supplier Spectrum Chemical Mfg Cor Supplier No. GL13625GMGL
Only null left

Chemical Identifiers

CAS 56-85-9
Molecular Formula C5H10N2O3
Molecular Weight (g/mol) 146.15
InChI Key ZDXPYRJPNDTMRX-VKHMYHEASA-N
IUPAC Name (2S)-2-amino-4-carbamoylbutanoic acid
SMILES N[C@@H](CCC(N)=O)C(O)=O

Specifications

CAS Max % 1
Loss on Drying 0.002
Packaging Amber Glass Bottle
Quantity 25 g
Specific Rotation +6.3° to +7.3°
Percent Purity 98.5 to 101.5%
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.