missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic Acid 99.0+%, TCI America™

Catalog No. G005925G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G005925G 25 g
G0059500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G005925G Supplier TCI America Supplier No. G005925G

Chemical Identifiers

CAS 56-86-0
Molecular Formula C5H9NO4
Molecular Weight (g/mol) 147.13
MDL Number MFCD00002634
InChI Key WHUUTDBJXJRKMK-UHFFFAOYNA-N
Synonym l-glutamic acid, glutamic acid, 2s-2-aminopentanedioic acid, h-glu-oh, s-2-aminopentanedioic acid, glutaminol, l-glutaminic acid, glutacid, glutamicol, glutamidex
PubChem CID 33032
ChEBI CHEBI:16015
IUPAC Name 2-aminopentanedioic acid
SMILES NC(CCC(O)=O)C(O)=O

Specifications

Melting Point 225°C
Color White
Quantity 25 g
Formula Weight 147.13
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Glutamic Acid
EINECSNumber (9)-1573
RTECSNumber LZ9700000
TSCA Yes