missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic acid monosodium salt monohydrate, 98+%

CAS: 6106-04-3 | C5H10NNaO5 | 187.13 g/mol

$61.26 - $379.53

Chemical Identifiers

CAS 6106-04-3
Molecular Formula C5H10NNaO5
Molecular Weight (g/mol) 187.13
MDL Number MFCD00150138
InChI Key GJBHGUUFMNITCI-QTNFYWBSSA-M
Synonym monosodium glutamate, l-glutamic acid monosodium salt monohydrate, p.a, l-glutamic acid monosodium salt monohydrate nt, l-glutamic acid monosodium salt monohydrate, saj special grade, monosodium glutamate, united states pharmacopeia usp reference standard, l-glutamic acid, monosodium salt, low endotoxin, pharmagrade, manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production., suitable for cell culture
PubChem CID 87090819
IUPAC Name (2S)-2-aminopentanedioic acid;sodium;hydrate
SMILES C(CC(=O)O)C(C(=O)O)N.O.[Na]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1291930
View Documents
Thermo Scientific Chemicals
A1291930
250 g
Each for $61.26
Only null left
 
AAA129190B
View Documents
Thermo Scientific Chemicals
A129190B
1000 g
Each for $109.29
Only null left
 
AAA129190I
View Documents
Thermo Scientific Chemicals
A129190I
5000 g
Each for $379.53
Only null left
 
Description

Description

L-Glutamic acid monosodium salt monohydrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals. Also used to intensify flavors of foods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Glutamic acid monosodium salt monohydrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals. Also used to intensify flavors of foods.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents
Specifications

Chemical Identifiers

6106-04-3
187.13
GJBHGUUFMNITCI-QTNFYWBSSA-M
87090819
C(CC(=O)O)C(C(=O)O)N.O.[Na]
C5H10NNaO5
MFCD00150138
monosodium glutamate, l-glutamic acid monosodium salt monohydrate, p.a, l-glutamic acid monosodium salt monohydrate nt, l-glutamic acid monosodium salt monohydrate, saj special grade, monosodium glutamate, united states pharmacopeia usp reference standard, l-glutamic acid, monosodium salt, low endotoxin, pharmagrade, manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production., suitable for cell culture
(2S)-2-aminopentanedioic acid;sodium;hydrate

Specifications

228°C (decomposition)
C5H10NNaO5
4164348
Soluble in water.
C(CC(=O)O)C(C(=O)O)N.O.[Na]
250 g
87090819
187.14 (169.12 Anhydrous)
L-Glutamic acid monosodium salt monohydrate
6106-04-3
MFCD00150138
monosodium glutamate, l-glutamic acid monosodium salt monohydrate, p.a, l-glutamic acid monosodium salt monohydrate nt, l-glutamic acid monosodium salt monohydrate, saj special grade, monosodium glutamate, united states pharmacopeia usp reference standard, l-glutamic acid, monosodium salt, low endotoxin, pharmagrade, manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production., suitable for cell culture
GJBHGUUFMNITCI-QTNFYWBSSA-M
(2S)-2-aminopentanedioic acid;sodium;hydrate
187.13
+22° (c=1 in 6N HCl)
≥98%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 205-538-1

RTECSNumber : MA1578000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only