missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(+)-Glutamic acid hydrochloride, 99%

Catalog No. AC165231000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC165231000 100 g Glass Bottle
AC165235000 500 g Glass Bottle
AC165230025 2.5 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC165231000 Supplier Thermo Scientific Chemicals Supplier No. 165231000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 138-15-8 | C5H10ClNO4 | 183.588 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 138-15-8
Molecular Formula C5H10ClNO4
Molecular Weight (g/mol) 183.588
MDL Number MFCD00012619
InChI Key RPAJSBKBKSSMLJ-DFWYDOINSA-N
Synonym l-glutamic acid hydrochloride, s-2-aminopentanedioic acid hydrochloride, l-+-glutamic acid hydrochloride, glutamic acid hydrochloride, unii-m0c2sp444t, glu hcl, acidogen, acidulin, gastuloric, glutamidin
PubChem CID 2723891
IUPAC Name (2S)-2-aminopentanedioic acid;hydrochloride
SMILES C(CC(=O)O)C(C(=O)O)N.Cl

Specifications

Ammonium 200ppm max.
Arsenic (As) 1ppm max.
pH 1 to 2.5 (2.5% soln. at 25°C)
Color White
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 10ppm max.
Loss on Drying 0.5% max. (105°C, 5 hrs)
Assay Percent Range 99%
Other Amino Acids 0.5% max. (TLC)
Packaging Glass Bottle
Linear Formula HO2CCH2CH2CH(NH2)CO2H·HCl
Beilstein 04, 488
Specific Rotation +25.2° to +25.8° (20°C, 589nm) (c=10, 2N HCl)
Sulfate 200ppm max.
Merck Index 15, 4505
Transmittance 95% min. (c=10, H2O, 430nm) 1cm cell
Sulfated Ash 0.1% max.
Specific Rotation Condition +25.50° (20°C c=10,2N HCl)
Quantity 100 g
Formula Weight 183.59
Percent Purity 98.5 to 101.5%
Physical Form Fine Crystalline Powder or Crystals
Chemical Name or Material L(+)-Glutamic acid hydrochloride, 99%
Show More Show Less
Hazard Category HYGROSCOPIC
Hazard Statement GHS P Statement
Causes serious eye damage.
Precautionary Statement GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 205-315-9
TSCA TSCA

RUO – Research Use Only