Learn More
Thermo Scientific™ L(+)-Glutamic acid hydrochloride, 99%
Supplier: Thermo Scientific™ 165230025
| Quantity | 2.5kg |
|---|---|
| Packaging | Plastic Bottle |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C5H10ClNO4 | |
| MFCD00012619 | |
| l-glutamic acid hydrochloride, s-2-aminopentanedioic acid hydrochloride, l-+-glutamic acid hydrochloride, glutamic acid hydrochloride, unii-m0c2sp444t, glu hcl, acidogen, acidulin, gastuloric, glutamidin | |
| (2S)-2-aminopentanedioic acid;hydrochloride |
Specifications
| L(+)-Glutamic acid hydrochloride | |
| 200ppm max. | |
| 1ppm max. | |
| Authentic | |
| 0.5% max. (105°C, 5 hrs) | |
| 0.5% max. (TLC) | |
| C5H10ClNO4 | |
| MFCD00012619 | |
| +25.2° to +25.8° (20°C, 589nm) (c=10, 2N HCl) | |
| 15, 4505 | |
| l-glutamic acid hydrochloride, s-2-aminopentanedioic acid hydrochloride, l-+-glutamic acid hydrochloride, glutamic acid hydrochloride, unii-m0c2sp444t, glu hcl, acidogen, acidulin, gastuloric, glutamidin | |
| RPAJSBKBKSSMLJ-DFWYDOINSA-N | |
| (2S)-2-aminopentanedioic acid;hydrochloride | |
| 183.588 | |
| 2723891 | |
| 98.5 to 101.5% |
| 99% | |
| 138-15-8 | |
| 10ppm max. | |
| 10ppm max. | |
| 99% | |
| Plastic Bottle | |
| HO2CCH2CH2CH(NH2)CO2H·HCl | |
| 04, 488 | |
| 200ppm max. | |
| 95% min. (c=10, H2O, 430nm) 1cm cell | |
| 0.1% max. | |
| C(CC(=O)O)C(C(=O)O)N.Cl | |
| +25.50° (20°C c=10,2N HCl) | |
| 2.5kg | |
| 183.59 |
Safety and Handling
GHS P Statement
Causes serious eye damage.
GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
HYGROSCOPIC
EINECSNumber : 205-315-9
TSCA : TSCA