missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic acid dimethyl ester hydrochloride, 98+%

Catalog No. AAL0376414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0376414 25 g
AAL0376406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0376414 Supplier Thermo Scientific Chemicals Supplier No. L0376414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 23150-65-4 | C7H13ClNO4 | 210.63 g/mol

L-Glutamic acid dimethyl ester hydrochloride is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Glutamic acid dimethyl ester hydrochloride is used as a pharmaceutical intermediate.

Solubility
Soluble in water, methanol (50 mg/ml).

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, water/ moisture, heat.

Chemical Identifiers

CAS 23150-65-4
Molecular Formula C7H13ClNO4
Molecular Weight (g/mol) 210.63
MDL Number MFCD00038879
InChI Key BPHCSYSXOKTCMA-UHFFFAOYNA-N
Synonym l-glutamic acid dimethyl ester hydrochloride, h-glu ome-ome.hcl, dimethyl l-glutamate hydrochloride, s-dimethyl 2-aminopentanedioate hydrochloride, unii-185vc5x3pf, 1,5-dimethyl 2s-2-aminopentanedioate hydrochloride, h-glu ome-ome inverted exclamation mark currencyhcl, s-dimethyl 2-aminopentanedioate hcl, glutamic acid dimethyl ester hydrochloride, dimethyl glutamic acid hydrochloride
PubChem CID 12917568
IUPAC Name dimethyl (2S)-2-aminopentanedioate;hydrochloride
SMILES [Cl].COC(=O)CCC(N)C(=O)OC

Specifications

Melting Point 99°C to 103°C
Beilstein 4238829
Solubility Information Soluble in water,methanol (50mg/ml).
Quantity 25 g
Odor Odorless
Formula Weight 211.65
Optical Rotation +26° (c=5 in Water)
Percent Purity ≥98%
Sensitivity Hygroscopic
Chemical Name or Material L-Glutamic acid dimethyl ester hydrochloride
EINECSNumber 245-461-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only