missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic acid di-tert-butyl ester hydrochloride, 98%

CAS: 32677-01-3 | C13H26ClNO4 | 295.804 g/mol

Supplier:  Thermo Scientific Chemicals H6370806

Quantity 5 g

 View more versions of this product

Catalog No. AAH6370806


Only null left

Description

Description

L-Glutamic acid di-tert-butyl ester may be useful in the synthesis of substance P antagonistic peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Glutamic acid di-tert-butyl ester may be useful in the synthesis of substance P antagonistic peptides.

Solubility
Soluble in methanol (50gmg/ml).

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
Specifications

Chemical Identifiers

32677-01-3
295.804
LFEYMWCCUAOUKZ-FVGYRXGTSA-N
208636
CC(C)(C)OC(=O)CCC(C(=O)OC(C)(C)C)N.Cl
C13H26ClNO4
MFCD00058003
l-glutamic acid di-tert-butyl ester hydrochloride, h-glu otbu-otbu.hcl, s-di-tert-butyl 2-aminopentanedioate hydrochloride, h-glu otbu-otbu hcl, l-glutamic acid, bis 1,1-dimethylethyl ester, hydrochloride, h-glu otbu-otbu inverted exclamation mark currencyhcl, glutamic acid di-t-butyl ester hydrochloride, s-di-tert-butyl 2-aminopentanedioate hcl, ditert-butyl 2s-2-aminopentanedioate hydrochloride, 1,5-di-tert-butyl 2s-2-aminopentanedioate hydrochloride
ditert-butyl (2S)-2-aminopentanedioate;hydrochloride

Specifications

L-Glutamic acid di-tert-butyl ester hydrochloride
C13H26ClNO4
3573738
Soluble in methanol (50gmg/ml).
CC(C)(C)OC(=O)CCC(C(=O)OC(C)(C)C)N.Cl
5 g
208636
98%
32677-01-3
MFCD00058003
l-glutamic acid di-tert-butyl ester hydrochloride, h-glu otbu-otbu.hcl, s-di-tert-butyl 2-aminopentanedioate hydrochloride, h-glu otbu-otbu hcl, l-glutamic acid, bis 1,1-dimethylethyl ester, hydrochloride, h-glu otbu-otbu inverted exclamation mark currencyhcl, glutamic acid di-t-butyl ester hydrochloride, s-di-tert-butyl 2-aminopentanedioate hcl, ditert-butyl 2s-2-aminopentanedioate hydrochloride, 1,5-di-tert-butyl 2s-2-aminopentanedioate hydrochloride
LFEYMWCCUAOUKZ-FVGYRXGTSA-N
ditert-butyl (2S)-2-aminopentanedioate;hydrochloride
295.804
295.8
Solid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : MA0880000

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions