missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic acid di-tert-butyl ester hydrochloride, 98%

Catalog No. AAH6370803
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6370803 1 g
AAH6370806 5 g
AAH6370814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6370803 Supplier Thermo Scientific Chemicals Supplier No. H6370803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 32677-01-3 | C13H26ClNO4 | 295.804 g/mol

L-Glutamic acid di-tert-butyl ester may be useful in the synthesis of substance P antagonistic peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Glutamic acid di-tert-butyl ester may be useful in the synthesis of substance P antagonistic peptides.

Solubility
Soluble in methanol (50gmg/ml).

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.

Chemical Identifiers

CAS 32677-01-3
Molecular Formula C13H26ClNO4
Molecular Weight (g/mol) 295.804
MDL Number MFCD00058003
InChI Key LFEYMWCCUAOUKZ-FVGYRXGTSA-N
Synonym l-glutamic acid di-tert-butyl ester hydrochloride, h-glu otbu-otbu.hcl, s-di-tert-butyl 2-aminopentanedioate hydrochloride, h-glu otbu-otbu hcl, l-glutamic acid, bis 1,1-dimethylethyl ester, hydrochloride, h-glu otbu-otbu inverted exclamation mark currencyhcl, glutamic acid di-t-butyl ester hydrochloride, s-di-tert-butyl 2-aminopentanedioate hcl, ditert-butyl 2s-2-aminopentanedioate hydrochloride, 1,5-di-tert-butyl 2s-2-aminopentanedioate hydrochloride
PubChem CID 208636
IUPAC Name ditert-butyl (2S)-2-aminopentanedioate;hydrochloride
SMILES CC(C)(C)OC(=O)CCC(C(=O)OC(C)(C)C)N.Cl

Specifications

Beilstein 3573738
Solubility Information Soluble in methanol (50gmg/ml).
Quantity 1 g
Formula Weight 295.8
Percent Purity 98%
Physical Form Solid
Chemical Name or Material L-Glutamic acid di-tert-butyl ester hydrochloride
RTECSNumber MA0880000
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only