missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic Acid, ≥99%, MP Biomedicals™

Catalog No. p-4679923
Change view
Click to view available options
Quantity:
100 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN1017931 100 g
ICN10179301 1 kg
ICN10179305 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN1017931 Supplier MP Biomedicals Inc Supplier No. 02101793.1
Only null left
Molecular biology, Cell Biology

Chemical Identifiers

CAS 56-86-0
Molecular Formula C5H9NO4
Molecular Weight (g/mol) 147.13
MDL Number MFCD00002634
InChI Key WHUUTDBJXJRKMK-UHFFFAOYNA-N
Synonym l-glutamic acid, glutamic acid, 2s-2-aminopentanedioic acid, h-glu-oh, s-2-aminopentanedioic acid, glutaminol, l-glutaminic acid, glutacid, glutamicol, glutamidex
PubChem CID 33032
ChEBI CHEBI:16015
SMILES NC(CCC(O)=O)C(O)=O

Specifications

Color White
Assay Percent Range ≥99%
Density 1.538g/mL at 20°C (Lit.)
IUPAC Name 2-aminopentanedioic acid
Quantity 100 g
Formula Weight 147.1
Physical Form Powder
Chemical Name or Material L-Glutamic Acid
EINECSNumber 200-293-7
Recommended Storage Store at room temperature (15° to 30°C).