missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Glutamic acid, 99%

CAS: 56-86-0 | C5H9NO4 | 147.13 g/mol

Supplier:  Thermo Scientific Chemicals A150310I

 View more versions of this product

Catalog No. AAA150310I


Only null left

Description

Description

It is used as a chiral building-block in the synthesis of the (S)-isomer of -butyrolactone-4-carboxylic acid.  L-Glutamic acid is widely used in fields of medicine, food processing, industry, etc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a chiral building-block in the synthesis of the (S)-isomer of -butyrolactone-4-carboxylic acid.  L-Glutamic acid is widely used in fields of medicine, food processing, industry, etc.

Solubility
It is very slightly soluble in cold water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under recommended storage conditions.
Specifications

Specifications

189°C to 192°C (decomposition)
56-86-0
C5H9NO4
1723801
l-glutamic acid, glutamic acid, 2s-2-aminopentanedioic acid, h-glu-oh, s-2-aminopentanedioic acid, glutaminol, l-glutaminic acid, glutacid, glutamicol, glutamidex
WHUUTDBJXJRKMK-UHFFFAOYNA-N
NC(CCC(O)=O)C(O)=O
147.13
33032
+31° (c=5 in 5N HCl)
≥99%
L-Glutamic acid
≥99%
MFCD00002634
14,4469
It is very slightly soluble in cold water.
1.525
(2S)-2-aminopentanedioic acid
5000 g
CHEBI:16015
147.13

Chemical Identifiers

56-86-0
147.13
WHUUTDBJXJRKMK-UHFFFAOYNA-N
33032
(2S)-2-aminopentanedioic acid
C5H9NO4
MFCD00002634
l-glutamic acid, glutamic acid, 2s-2-aminopentanedioic acid, h-glu-oh, s-2-aminopentanedioic acid, glutaminol, l-glutaminic acid, glutacid, glutamicol, glutamidex
CHEBI:16015
NC(CCC(O)=O)C(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H500

EINECSNumber : 200-293-7

RTECSNumber : LZ9700000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only