missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(+)-Glutamic acid, 99%

Numéro de catalogue. AC156212500
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
5 kg
Packaging:
Plastic Bottle
Plastic Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
AC156212500 250 g Plastic Bottle
AC156211000 100 g Plastic Bottle
AC156210010 1 kg Plastic Bottle
AC156210050 5 kg Plastic Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Numéro de catalogue. AC156212500 Fournisseur Thermo Scientific Chemicals Code fournisseur 156212500
Il en reste null

CAS: 56-86-0 | C5H9NO4 | 147.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 56-86-0
Molecular Formula C5H9NO4
Molecular Weight (g/mol) 147.13
MDL Number MFCD00002634
InChI Key WHUUTDBJXJRKMK-UHFFFAOYNA-N
Synonym l-glutamic acid, glutamic acid, 2s-2-aminopentanedioic acid, h-glu-oh, s-2-aminopentanedioic acid, glutaminol, l-glutaminic acid, glutacid, glutamicol, glutamidex
PubChem CID 33032
ChEBI CHEBI:16015
IUPAC Name (2S)-2-aminopentanedioic acid
SMILES NC(CCC(O)=O)C(O)=O

Spécifications

Ammonium 200ppm max.
Color White
Chlorine (Cl) 200ppm max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 30ppm max.
Loss on Drying 0.5% (105°C, 3 hrs) max.
Assay Percent Range 99%
Other Amino Acids Not detectable
Packaging Plastic Bottle
Linear Formula HO2CCH2CH2CH(NH2)CO2H
Specific Rotation +30.5° to +32.5° (20°C, 589nm) (on dried substance) (c=10, 2N HCl)
Sulfate 300ppm max.
Sulfated Ash 0.1% max.
Sublimation Point >200
Specific Rotation Condition +32.00 (20.00°C c=10,2N HCl)
Quantity 250 g
Formula Weight 147.13
Percent Purity 98.5 to 100.5% (on dried substance)
Physical Form Powder
Chemical Name or Material L(+)-Glutamic acid
Afficher plus de résultats Afficher moins de résultats

RUO – Research Use Only