missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(+)-Glutamic acid, 99%

CAS: 56-86-0 | C5H9NO4 | 147.13 g/mol

Supplier:  Thermo Scientific Chemicals 156210010

Quantity 1 kg
Packaging Plastic Bottle

 View more versions of this product

Catalog No. AC156210010


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

56-86-0
147.13
WHUUTDBJXJRKMK-UHFFFAOYNA-N
33032
(2S)-2-aminopentanedioic acid
C5H9NO4
MFCD00002634
l-glutamic acid, glutamic acid, 2s-2-aminopentanedioic acid, h-glu-oh, s-2-aminopentanedioic acid, glutaminol, l-glutaminic acid, glutacid, glutamicol, glutamidex
CHEBI:16015
NC(CCC(O)=O)C(O)=O

Specifications

L(+)-Glutamic acid
56-86-0
200ppm max.
Authentic
0.5% (105°C, 3 hrs) max.
Not detectable
C5H9NO4
MFCD00002634
300ppm max.
0.1% max.
WHUUTDBJXJRKMK-UHFFFAOYNA-N
+32.00 (20.00°C c=10,2N HCl)
1 kg
33032
147.13
Powder
200ppm max.
White
10ppm max.
30ppm max.
99%
Plastic Bottle
HO2CCH2CH2CH(NH2)CO2H
+30.5° to +32.5° (20°C, 589nm) (on dried substance) (c=10, 2N HCl)
l-glutamic acid, glutamic acid, 2s-2-aminopentanedioic acid, h-glu-oh, s-2-aminopentanedioic acid, glutaminol, l-glutaminic acid, glutacid, glutamicol, glutamidex
>200
NC(CCC(O)=O)C(O)=O
(2S)-2-aminopentanedioic acid
147.13
CHEBI:16015
98.5 to 100.5% (on dried substance)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions