missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Glutamic Acid, 98.5-101.5%, Spectrum™ Chemical
GL125, 56-86-0, C5H9NO4
Supplier: Spectrum Chemical Mfg Cor GL1251KG
Description
Spectrum™ Chemical L-Glutamic Acid , also known as sodium glutamate, is the sodium salt of glutamic acid. It is used in the food industry most usually as a flavor with an umami taste. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.Specifications
| 56-86-0 | |
| 0.2% | |
| C5H9NO4 | |
| 1 kg | |
| WHUUTDBJXJRKMK-UHFFFAOYNA-N | |
| 2-aminopentanedioic acid | |
| 98.5 to 101.5% |
| 100% | |
| Poly Pail | |
| MFCD00002634 | |
| +31.5° to +32.2° | |
| NC(CCC(O)=O)C(O)=O | |
| 147.13 | |
| Reagent |
Chemical Identifiers
| 56-86-0 | |
| 147.13 | |
| WHUUTDBJXJRKMK-UHFFFAOYNA-N | |
| NC(CCC(O)=O)C(O)=O |
| C5H9NO4 | |
| MFCD00002634 | |
| 2-aminopentanedioic acid |