missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic Acid, 98.5-101.5%, Spectrum™ Chemical

Catalog No. 18603820
Change view
Click to view available options
Quantity:
100 g
1 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603820 100 g Poly Bottle
18603821 1 kg Poly Pail
18603822 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18603820 Supplier Spectrum Chemical Mfg Cor Supplier No. GL125100GM
Only null left

GL125, 56-86-0, C5H9NO4

Spectrum™ Chemical L-Glutamic Acid , also known as sodium glutamate, is the sodium salt of glutamic acid. It is used in the food industry most usually as a flavor with an umami taste. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 56-86-0
Molecular Formula C5H9NO4
Molecular Weight (g/mol) 147.13
MDL Number MFCD00002634
InChI Key WHUUTDBJXJRKMK-UHFFFAOYNA-N
IUPAC Name 2-aminopentanedioic acid
SMILES NC(CCC(O)=O)C(O)=O

Specifications

CAS Max % 100%
Loss on Drying 0.2%
Packaging Poly Bottle
Quantity 100 g
Specific Rotation +31.5° to +32.2°
Percent Purity 98.5 to 101.5%
Grade Reagent