missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic acid 5-tert-butyl ester, 98%, Thermo Scientific Chemicals

Catalog No. AAH6322306
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6322306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6322306 Supplier Thermo Scientific Chemicals Supplier No. H6322306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

L-Glutamic acid 5-tert-butyl ester is used as a fine chemical intermediate, pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Glutamic acid 5-tert-butyl ester is used as a fine chemical intermediate, pharmaceutical intermediate.

Solubility
Sparingly soluble in water; practically insoluble in ethanol or ether.

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 2419-56-9
Molecular Formula C9H17NO4
Molecular Weight (g/mol) 203.238
MDL Number MFCD00038580
InChI Key OIOAKXPMBIZAHL-LURJTMIESA-N
PubChem CID 7010573
IUPAC Name (2S)-2-amino-5-[(2-methylpropan-2-yl)oxy]-5-oxopentanoic acid
SMILES CC(C)(C)OC(=O)CCC(C(=O)O)N

Specifications

Beilstein 2049031
Synonym h-glu otbu-oh, l-glutamic acid 5-tert-butyl ester, h-glu o-t-bu-oh, 2s-2-amino-5-tert-butoxy-5-oxopentanoic acid, l-gamma-t-butyl glutamate, l-glutamic acid,5-1,1-dimethylethyl ester, 5-tert-butyl l-glutamate, l-glutamic acid, 5-1,1-dimethylethyl ester, glu otbu, glu tbu
Solubility Information Sparingly soluble in water; practically insoluble in ethanol or ether.
Quantity 5 g
Formula Weight 203.24
Percent Purity 98%
Physical Form Solid
Chemical Name or Material L-Glutamic acid 5-tert-butyl ester
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only