missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic acid 5-tert-butyl 1-methyl ester hydrochloride, 98%

CAS: 6234-01-1 | C10H20ClNO4 | 253.72 g/mol

$133.34 - $485.73

Chemical Identifiers

CAS 6234-01-1
Molecular Formula C10H20ClNO4
Molecular Weight (g/mol) 253.72
MDL Number MFCD00153438
InChI Key YIFPACFSZQWAQF-RZUXQYDTNA-N
Synonym h-glu otbu-ome.hcl, l-glutamic acid 5-tert-butyl 1-methyl ester hydrochloride, h-glu otbu-ome hcl, 5-tert-butyl 1-methyl l-glutamate hydrochloride, h-glu otbu-ome inverted exclamation mark currencyhcl, h-glu otbu-ome hydrochloride, s-5-tert-butyl 1-methyl 2-aminopentanedioate hydrochloride, h-glu otbu-omehcl, 5-tert-butyl 1-methyl 2s-2-aminopentanedioate hydrochloride, glu otbu-ome.hcl
PubChem CID 16218751
IUPAC Name 5-O-tert-butyl 1-O-methyl (2S)-2-aminopentanedioate;hydrochloride
SMILES CC(C)(C)OC(=O)CCC(C(=O)OC)N.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6366103
View Documents
Thermo Scientific Chemicals
H6366103
1 g
Each for $133.34
Only null left
 
AAH6366106
View Documents
Thermo Scientific Chemicals
H6366106
5 g
Each for $485.73
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6234-01-1
253.72
YIFPACFSZQWAQF-RZUXQYDTNA-N
16218751
CC(C)(C)OC(=O)CCC(C(=O)OC)N.Cl
C10H20ClNO4
MFCD00153438
h-glu otbu-ome.hcl, l-glutamic acid 5-tert-butyl 1-methyl ester hydrochloride, h-glu otbu-ome hcl, 5-tert-butyl 1-methyl l-glutamate hydrochloride, h-glu otbu-ome inverted exclamation mark currencyhcl, h-glu otbu-ome hydrochloride, s-5-tert-butyl 1-methyl 2-aminopentanedioate hydrochloride, h-glu otbu-omehcl, 5-tert-butyl 1-methyl 2s-2-aminopentanedioate hydrochloride, glu otbu-ome.hcl
5-O-tert-butyl 1-O-methyl (2S)-2-aminopentanedioate;hydrochloride

Specifications

6234-01-1
MFCD00153438
h-glu otbu-ome.hcl, l-glutamic acid 5-tert-butyl 1-methyl ester hydrochloride, h-glu otbu-ome hcl, 5-tert-butyl 1-methyl l-glutamate hydrochloride, h-glu otbu-ome inverted exclamation mark currencyhcl, h-glu otbu-ome hydrochloride, s-5-tert-butyl 1-methyl 2-aminopentanedioate hydrochloride, h-glu otbu-omehcl, 5-tert-butyl 1-methyl 2s-2-aminopentanedioate hydrochloride, glu otbu-ome.hcl
CC(C)(C)OC(=O)CCC(C(=O)OC)N.Cl
1 g
16218751
98%
L-Glutamic acid 5-tert-butyl 1-methyl ester hydrochloride
C10H20ClNO4
3572140
YIFPACFSZQWAQF-RZUXQYDTNA-N
5-O-tert-butyl 1-O-methyl (2S)-2-aminopentanedioate;hydrochloride
253.72
253.72
Solid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 228-348-0

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only