missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic acid 5-tert-butyl 1-methyl ester hydrochloride, 98%

Catalog No. AAH6366103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6366103 1 g
AAH6366106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6366103 Supplier Thermo Scientific Chemicals Supplier No. H6366103

CAS: 6234-01-1 | C10H20ClNO4 | 253.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6234-01-1
Molecular Formula C10H20ClNO4
Molecular Weight (g/mol) 253.72
MDL Number MFCD00153438
InChI Key YIFPACFSZQWAQF-RZUXQYDTNA-N
Synonym h-glu otbu-ome.hcl, l-glutamic acid 5-tert-butyl 1-methyl ester hydrochloride, h-glu otbu-ome hcl, 5-tert-butyl 1-methyl l-glutamate hydrochloride, h-glu otbu-ome inverted exclamation mark currencyhcl, h-glu otbu-ome hydrochloride, s-5-tert-butyl 1-methyl 2-aminopentanedioate hydrochloride, h-glu otbu-omehcl, 5-tert-butyl 1-methyl 2s-2-aminopentanedioate hydrochloride, glu otbu-ome.hcl
PubChem CID 16218751
IUPAC Name 5-O-tert-butyl 1-O-methyl (2S)-2-aminopentanedioate;hydrochloride
SMILES CC(C)(C)OC(=O)CCC(C(=O)OC)N.Cl

Specifications

Beilstein 3572140
Quantity 1 g
Formula Weight 253.72
Percent Purity 98%
Physical Form Solid
Chemical Name or Material L-Glutamic acid 5-tert-butyl 1-methyl ester hydrochloride
EINECSNumber 228-348-0
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only