missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Glutamic acid 5-methyl ester, 99%

CAS: 1499-55-4 | C6H11NO4 | 161.157 g/mol

Supplier:  Thermo Scientific Chemicals B2485922

 View more versions of this product

Catalog No. AAB2485922


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

182°C (decomposition)
1499-55-4
MFCD00002632
ZGEYCCHDTIDZAE-BYPYZUCNSA-N
(2S)-2-amino-5-methoxy-5-oxopentanoic acid
161.157
161.16
L-Glutamic acid 5-methyl ester
C6H11NO4
l-glutamic acid 5-methyl ester, h-glu ome-oh, polymethylglutamate, 2s-2-amino-5-methoxy-5-oxopentanoic acid, l-glutamic acid gamma-methyl ester, 5-methyl l-glutamate, l-glutamic acid, 5-methyl ester, 2s-2-amino-5-methoxy-5-oxo-pentanoic acid, 2s-2-amino-4-methoxycarbonyl butanoic acid
COC(=O)CCC(C(=O)O)N
100 g
68662
99%

Chemical Identifiers

1499-55-4
161.157
ZGEYCCHDTIDZAE-BYPYZUCNSA-N
68662
COC(=O)CCC(C(=O)O)N
C6H11NO4
MFCD00002632
l-glutamic acid 5-methyl ester, h-glu ome-oh, polymethylglutamate, 2s-2-amino-5-methoxy-5-oxopentanoic acid, l-glutamic acid gamma-methyl ester, 5-methyl l-glutamate, l-glutamic acid, 5-methyl ester, 2s-2-amino-5-methoxy-5-oxo-pentanoic acid, 2s-2-amino-4-methoxycarbonyl butanoic acid
(2S)-2-amino-5-methoxy-5-oxopentanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 216-110-9

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only