missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals L-Glutamic acid 5-methyl ester, 99%
CAS: 1499-55-4 | C6H11NO4 | 161.157 g/mol
Supplier: Thermo Scientific Chemicals B2485906
| Quantity | 5 g |
|---|
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C6H11NO4 | |
| MFCD00002632 | |
| l-glutamic acid 5-methyl ester, h-glu ome-oh, polymethylglutamate, 2s-2-amino-5-methoxy-5-oxopentanoic acid, l-glutamic acid gamma-methyl ester, 5-methyl l-glutamate, l-glutamic acid, 5-methyl ester, 2s-2-amino-5-methoxy-5-oxo-pentanoic acid, 2s-2-amino-4-methoxycarbonyl butanoic acid | |
| (2S)-2-amino-5-methoxy-5-oxopentanoic acid |
Specifications
| 182°C (decomposition) | |
| 1499-55-4 | |
| MFCD00002632 | |
| ZGEYCCHDTIDZAE-BYPYZUCNSA-N | |
| (2S)-2-amino-5-methoxy-5-oxopentanoic acid | |
| 161.157 | |
| 161.16 |
| L-Glutamic acid 5-methyl ester | |
| C6H11NO4 | |
| l-glutamic acid 5-methyl ester, h-glu ome-oh, polymethylglutamate, 2s-2-amino-5-methoxy-5-oxopentanoic acid, l-glutamic acid gamma-methyl ester, 5-methyl l-glutamate, l-glutamic acid, 5-methyl ester, 2s-2-amino-5-methoxy-5-oxo-pentanoic acid, 2s-2-amino-4-methoxycarbonyl butanoic acid | |
| COC(=O)CCC(C(=O)O)N | |
| 5 g | |
| 68662 | |
| 99% |
Safety and Handling
EINECSNumber : 216-110-9
TSCA : No
Recommended Storage : Keep cold