missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic acid 5-benzyl ester, 99%

Catalog No. AAB2218403
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2218403 1 g
AAB2218406 5 g
AAB2218414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2218403 Supplier Thermo Scientific Chemicals Supplier No. B2218403

CAS: 1676-73-9 | C12H15NO4 | 237.255 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1676-73-9
Molecular Formula C12H15NO4
Molecular Weight (g/mol) 237.255
MDL Number MFCD00002633
InChI Key BGGHCRNCRWQABU-JTQLQIEISA-N
Synonym h-glu obzl-oh, gamma-benzyl l-glutamate, l-glutamic acid 5-benzyl ester, 5-benzyl l-glutamate, l-glutamic acid gamma-benzyl ester, s-2-amino-5-benzyloxy-5-oxopentanoic acid, 2s-2-amino-5-benzyloxy-5-oxopentanoic acid, glutamic acid, 5-benzyl ester, l, gama-benzyl l-glutamate, h-glu obzl-oh.hcl
PubChem CID 122337
IUPAC Name (2S)-2-amino-5-oxo-5-phenylmethoxypentanoic acid
SMILES C1=CC=C(C=C1)COC(=O)CCC(C(=O)O)N

Specifications

Melting Point 162°C to 165°C
Beilstein 1885646
Quantity 1 g
Formula Weight 237.26
Optical Rotation +23° (c=1 in Acetic Acid)
Percent Purity 99%
Chemical Name or Material L-Glutamic acid 5-benzyl ester
EINECSNumber 216-826-1
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only