missing translation for 'onlineSavingsMsg'
Learn More

L-Dithiothreitol 95.0+%, TCI America™

Catalog No. D15895G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D15895G 5 g
D15891G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D15895G Supplier TCI America Supplier No. D15895G

Chemical Identifiers

CAS 16096-97-2
Molecular Formula C4H10O2S2
Molecular Weight (g/mol) 154.24
MDL Number MFCD00064305
InChI Key VHJLVAABSRFDPM-UHFFFAOYNA-N
Synonym l-dithiothreitol, l-dtt, dithiothreitol, 1,4-dithiothreitol, l-1,4-dithiothreitol, cleland reagent chiral, l---dithiothreitol, 2,3-dihydroxy-1,4-dithiobutane, 2r,3r-1,4-dimercapto-2,3-butanediol, rel-2r,3r-1,4-dimercaptobutane-2,3-diol
PubChem CID 439196
ChEBI CHEBI:42106
IUPAC Name 1,4-disulfanylbutane-2,3-diol
SMILES OC(CS)C(O)CS

Specifications

Melting Point 51°C
Color White
Quantity 5 g
Formula Weight 154.24
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Dithiothreitol
RTECSNumber EK1612000
TSCA No
Recommended Storage Freezer