missing translation for 'onlineSavingsMsg'
Learn More

L-Dihydroorotic acid, 99%

Catalog No. AC437240010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC437240010 1 g Glass bottle
AC437240050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC437240010 Supplier Thermo Scientific Chemicals Supplier No. 437240010

CAS: 5988-19-2 | C5H6N2O4 | 158.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5988-19-2
Molecular Formula C5H6N2O4
Molecular Weight (g/mol) 158.11
InChI Key UFIVEPVSAGBUSI-REOHCLBHSA-N
Synonym l-dihydroorotic acid, s-2,6-dioxohexahydropyrimidine-4-carboxylic acid, l-dihydroorotate, dihydro-l-orotic acid, s-dihydroorotic acid, l-hydroorotic acid, s-dihydroorotate, hydroorotic acid, l, 4s-2,6-dioxo-1,3-diazinane-4-carboxylic acid, s-4,5-dihydroorotate
PubChem CID 439216
ChEBI CHEBI:17025
IUPAC Name (4S)-2,6-dioxo-1,3-diazinane-4-carboxylic acid
SMILES C1C(NC(=O)NC1=O)C(=O)O

Specifications

Melting Point 254.0°C to 255.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 158.11
Percent Purity 99%
Physical Form Powder
Chemical Name or Material L-Dihydroorotic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:

RUO – Research Use Only