missing translation for 'onlineSavingsMsg'
Learn More

L-Dihydroorotic acid, 98%

Catalog No. AAH6042906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6042906 5 g
AAH6042903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6042906 Supplier Thermo Scientific Chemicals Supplier No. H6042906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5988-19-2 | C5H6N2O4 | 158.113 g/mol

L-Dihydroorotic acid a metabolite.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Dihydroorotic acid a metabolite.

Solubility
Soluble in water (partly), and dimethyl formamide.

Notes
Store at room temperature. Keep away from oxidizing agents.

Chemical Identifiers

CAS 5988-19-2
Molecular Formula C5H6N2O4
Molecular Weight (g/mol) 158.113
MDL Number MFCD00085339
InChI Key UFIVEPVSAGBUSI-REOHCLBHSA-N
Synonym l-dihydroorotic acid, s-2,6-dioxohexahydropyrimidine-4-carboxylic acid, l-dihydroorotate, dihydro-l-orotic acid, s-dihydroorotic acid, l-hydroorotic acid, s-dihydroorotate, hydroorotic acid, l, 4s-2,6-dioxo-1,3-diazinane-4-carboxylic acid, s-4,5-dihydroorotate
PubChem CID 439216
ChEBI CHEBI:17025
IUPAC Name (4S)-2,6-dioxo-1,3-diazinane-4-carboxylic acid
SMILES C1C(NC(=O)NC1=O)C(=O)O

Specifications

Melting Point 254°C to 255°C (decomposition)
Quantity 5 g
Solubility Information Soluble in water (partly),and dimethyl formamide.
Formula Weight 158.11
Percent Purity 98%
Chemical Name or Material L-Dihydroorotic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only