missing translation for 'onlineSavingsMsg'
Learn More

L-(-)-Cystine 98.0+%, TCI America™

Catalog No. C051925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C051925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C051925G Supplier TCI America Supplier No. C051925G

Specifications

Color White
Physical Form Crystal-Powder at 20°C
CAS 56-89-3
Melting Point 261°C
Molecular Formula C6H12N2O4S2
MDL Number MFCD00064228
Synonym l-cystine, cystine, l-dicysteine, l-cystin, cystine acid, cysteine disulfide, oxidized l-cysteine, beta,beta'-dithiodialanine, h-cys-oh 2, l-cysteine disulfide
Quantity 25 g
InChI Key LEVWYRKDKASIDU-IMJSIDKUSA-N
SMILES C(C(C(=O)O)N)SSCC(C(=O)O)N
IUPAC Name (2R)-2-amino-3-[[(2R)-2-amino-2-carboxyethyl]disulfanyl]propanoic acid
Molecular Weight (g/mol) 240.292
PubChem CID 67678
ChEBI CHEBI:16283
Formula Weight 240.29
Percent Purity ≥98.0% (HPLC,N)
Chemical Name or Material L-(-)-Cystine
Show More Show Less
EINECSNumber (9)-1587&(2)-1468
RTECSNumber HA2690000
TSCA Yes