missing translation for 'onlineSavingsMsg'
Learn More

L-(-)-Cystine Dihydrochloride 98.0+%, TCI America™

Catalog No. C052025G
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C052025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. C052025G missing translation for 'mfr' TCI America missing translation for 'supplierNo' C052025G

Chemical Identifiers

CAS 30925-07-6
Molecular Formula C6H14Cl2N2O4S2
Molecular Weight (g/mol) 313.208
MDL Number MFCD00070399
InChI Key HHGZUQPEIHGQST-RGVONZFCSA-N
Synonym l-cystine dihydrochloride, l---cystine dihydrochloride, cystine, dihydrochloride, h-cys-oh 2.2hcl, c6h12n2o4s2.2hcl, h-cys-oh 2 . 2hcl, l-cystine dihydrochloride tlc, l-cystine dihydrochloride, cell culture reagent, 2r-2-amino-3-2r-2-amino-2-carboxyethyl disulfanyl propanoic acid dihydrochloride, l-cystine dihydrochloride, from non-animal source, bioreagent, suitable for cell culture dry basis
PubChem CID 21121987
IUPAC Name (2R)-2-amino-3-[[(2R)-2-amino-2-carboxyethyl]disulfanyl]propanoic acid;dihydrochloride
SMILES C(C(C(=O)O)N)SSCC(C(=O)O)N.Cl.Cl

Specifications

Color White
Quantity 25 g
Formula Weight 313.21
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material L-(-)-Cystine Dihydrochloride