missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Cystine dihydrochloride, 99%
CAS: 30925-07-6 | C6H14Cl2N2O4S2 | 313.208 g/mol
$131.68 - $834.99
Chemical Identifiers
| CAS | 30925-07-6 |
|---|---|
| Molecular Formula | C6H14Cl2N2O4S2 |
| Molecular Weight (g/mol) | 313.208 |
| MDL Number | MFCD00070399 |
| InChI Key | HHGZUQPEIHGQST-RGVONZFCSA-N |
| Synonym | l-cystine dihydrochloride, l---cystine dihydrochloride, cystine, dihydrochloride, h-cys-oh 2.2hcl, c6h12n2o4s2.2hcl, h-cys-oh 2 . 2hcl, l-cystine dihydrochloride tlc, l-cystine dihydrochloride, cell culture reagent, 2r-2-amino-3-2r-2-amino-2-carboxyethyl disulfanyl propanoic acid dihydrochloride, l-cystine dihydrochloride, from non-animal source, bioreagent, suitable for cell culture dry basis |
| PubChem CID | 21121987 |
| IUPAC Name | (2R)-2-amino-3-[[(2R)-2-amino-2-carboxyethyl]disulfanyl]propanoic acid;dihydrochloride |
| SMILES | C(C(C(=O)O)N)SSCC(C(=O)O)N.Cl.Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ6229214
|
Thermo Scientific Chemicals
J6229214 |
25 g |
Each for $131.68
|
|
|||||
|
AAJ6229222
|
Thermo Scientific Chemicals
J6229222 |
100 g |
Each for $381.30
|
|
|||||
|
AAJ6229230
|
Thermo Scientific Chemicals
J6229230 |
250 g |
Each for $834.99
|
|
|||||
Description
Non-animal sourced L-cystine is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsNon-animal sourced L-cystine is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies.
Solubility
Soluble in hydrochloric acid. (0.5 M HCl: 50 mg/mL) Soluble in water.
Notes
Store at room temperature. Incompatible with oxidizing agents.
Chemical Identifiers
| 30925-07-6 | |
| 313.208 | |
| HHGZUQPEIHGQST-RGVONZFCSA-N | |
| 21121987 | |
| C(C(C(=O)O)N)SSCC(C(=O)O)N.Cl.Cl |
| C6H14Cl2N2O4S2 | |
| MFCD00070399 | |
| l-cystine dihydrochloride, l---cystine dihydrochloride, cystine, dihydrochloride, h-cys-oh 2.2hcl, c6h12n2o4s2.2hcl, h-cys-oh 2 . 2hcl, l-cystine dihydrochloride tlc, l-cystine dihydrochloride, cell culture reagent, 2r-2-amino-3-2r-2-amino-2-carboxyethyl disulfanyl propanoic acid dihydrochloride, l-cystine dihydrochloride, from non-animal source, bioreagent, suitable for cell culture dry basis | |
| (2R)-2-amino-3-[[(2R)-2-amino-2-carboxyethyl]disulfanyl]propanoic acid;dihydrochloride |
Specifications
| 30925-07-6 | |
| White to Yellow | |
| MFCD00070399 | |
| l-cystine dihydrochloride, l---cystine dihydrochloride, cystine, dihydrochloride, h-cys-oh 2.2hcl, c6h12n2o4s2.2hcl, h-cys-oh 2 . 2hcl, l-cystine dihydrochloride tlc, l-cystine dihydrochloride, cell culture reagent, 2r-2-amino-3-2r-2-amino-2-carboxyethyl disulfanyl propanoic acid dihydrochloride, l-cystine dihydrochloride, from non-animal source, bioreagent, suitable for cell culture dry basis | |
| HHGZUQPEIHGQST-RGVONZFCSA-N | |
| (2R)-2-amino-3-[[(2R)-2-amino-2-carboxyethyl]disulfanyl]propanoic acid;dihydrochloride | |
| 21121987 | |
| 99% | |
| L-Cystine dihydrochloride |
| 228°C to 232°C | |
| C6H14Cl2N2O4S2 | |
| 25 g | |
| Soluble in hydrochloric acid. (0.5 M HCl: 50mg/mL) Soluble in water. | |
| C(C(C(=O)O)N)SSCC(C(=O)O)N.Cl.Cl | |
| 313.208 | |
| 313.22 | |
| Powder |
Safety and Handling
EINECSNumber : 250-391-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only