missing translation for 'onlineSavingsMsg'
Learn More

L-Cystine dihydrochloride, 98%

Catalog No. AAJ63717A1
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63717A1 1 kg
AAJ6371730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63717A1 Supplier Thermo Scientific Chemicals Supplier No. J63717A1

CAS: 30925-07-6 | C6H14Cl2N2O4S2 | 313.208 g/mol

Non-animal sourced L-cystine is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies. L-cystine is used as a cell culture supplement.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Non-animal sourced L-cystine is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies. L-cystine is used as a cell culture supplement.

Solubility
Freely soluble in Dilute hydrochloric acid.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 30925-07-6
Molecular Formula C6H14Cl2N2O4S2
Molecular Weight (g/mol) 313.208
MDL Number MFCD00070399
InChI Key HHGZUQPEIHGQST-RGVONZFCSA-N
Synonym l-cystine dihydrochloride, l---cystine dihydrochloride, cystine, dihydrochloride, h-cys-oh 2.2hcl, c6h12n2o4s2.2hcl, h-cys-oh 2 . 2hcl, l-cystine dihydrochloride tlc, l-cystine dihydrochloride, cell culture reagent, 2r-2-amino-3-2r-2-amino-2-carboxyethyl disulfanyl propanoic acid dihydrochloride, l-cystine dihydrochloride, from non-animal source, bioreagent, suitable for cell culture dry basis
PubChem CID 21121987
IUPAC Name (2R)-2-amino-3-[[(2R)-2-amino-2-carboxyethyl]disulfanyl]propanoic acid;dihydrochloride
SMILES C(C(C(=O)O)N)SSCC(C(=O)O)N.Cl.Cl

Specifications

Melting Point 228°C to 232°C
Color White to Yellow
Solubility Information Freely soluble in Dilute hydrochloric acid.
Quantity 1 kg
Formula Weight 313.22
Grade Cell Culture Reagent
Physical Form Powder
Chemical Name or Material L-Cystine dihydrochloride
EINECSNumber 250-391-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only