missing translation for 'onlineSavingsMsg'
Learn More

L-(-)-Cystine, BAKER ANALYZED™ Biochemical Reagent, J.T. Baker™

Catalog No. 02003291
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02003291 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02003291 Supplier Avantor Supplier No. G12305

High quality biochemicals for research use

Chemical Identifiers

CAS 56-89-3
Molecular Formula C6H12N2O4S2
Molecular Weight (g/mol) 240.292
InChI Key LEVWYRKDKASIDU-IMJSIDKUSA-N
Synonym l-cystine, cystine, l-dicysteine, l-cystin, cystine acid, cysteine disulfide, oxidized l-cysteine, beta,beta'-dithiodialanine, h-cys-oh 2, l-cysteine disulfide
PubChem CID 67678
ChEBI CHEBI:16283
IUPAC Name (2R)-2-amino-3-[[(2R)-2-amino-2-carboxyethyl]disulfanyl]propanoic acid
SMILES C(C(C(=O)O)N)SSCC(C(=O)O)N

Specifications

Packaging Poly Bottle
Quantity 100 g
Formula Weight 240.3
Grade BAKER ANALYZED™ Biochemical Reagent
Chemical Name or Material L-(-)-Cystine